Cole-Parmer® ESD Safe Plastic Tweezers
Handle sensitive electronic components safely
  • Chemical resistant to oils, grease, fuels, and nonpolar solvents
  • Withstand heat to 226°F (130°C)
ESD-safe plastic tweezers, reinforced with 30% carbon fiber, offer excellent wear and abrasion resistance. Featuring hard rigidity, high tensile and flexural strength, these tweezers are ideal for handling sensitive materials such as electronic components, micro-mechanical parts, glass, and ceramic substrates.
1,200.00 - ₹10,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
ESD Safe Plastic Tweezer, Tips; Flat, Duckbill, 11.5 cm
₹1,200.00INR / Each
COMPARE
ESD Safe Plastic Tweezer, Tips; Fine, Sharp, 11.5 cm
₹1,200.00INR / Each
COMPARE
ESD Safe Plastic Tweezer, Tips; Angled, Pointed, 11.5 cm
₹1,200.00INR / Each
COMPARE
ESD Safe Plastic Tweezer, Tips; Curved, Angled, Pointed, 11.5 cm
₹1,200.00INR / Each
COMPARE
ESD Safe Plastic Tweezer, Tips; Bent, Angled, Pointed, 11.5 cm
₹1,200.00INR / Each
COMPARE
ESD Safe Plastic Tweezer, Tips; Angled, Squared, Flat, Trapezoid, 11.5 cm
₹1,200.00INR / Each
COMPARE
Full Plastic Tweezer Set w/ Hard Case
₹10,000.00INR / Set of 5
Showing 1- 7 of 7