Cole-Parmer® Fluoride-Resistant, Self-Cleaning Cartridge pH Electrodes
Keep original housing and simply replace the electrode cartridge
  • Fluid flow across the surface provides a cleaning action that extends the life of the electrode and improves performance
  • Flat measuring surface minimizes electrode coating, scratching, and breakage
These electrodes are ideal for solutions containing acidic fluorides (hydrofluoric acid).
89,900.00 - ₹118,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
HF Resistant Submersible pH Electrode, CPVC
₹89,900.00INR / Each
COMPARE
HF Resistant Submersible pH Electrode, CPVC, 100 Ohms RTD
₹118,000.00INR / Each
COMPARE
HF Resistant Submersible pH Electrode, CPVC, 1 k Ohms RTD
₹118,000.00INR / Each