Cole-Parmer® FSB-200 Series Temperature Controller for Sand Bath Systems
Designed to improve the temperature stability and accuracy of FSB-200 and FSB-200D fluidized sand baths
  • Precision PID controller with type K thermocouple probe
  • Fast heat-ups and accurate temperature control to 630 °C (1166 °F)
  • Built-in flowmeter for air flow control
Temperature controller with integrated flowmeter and PID temperature controller with dual-setpoint programming improves temperature accuracy and stability. Easily access two programmable setpoints for faster operation. A single-point calibration feature ensures accurate temperature readings. Set and view temperature in °C or °F to 1° resolution. The type K thermocouple probe (included) fits into the thermowell of the bath for accurate measurements.
355,000.00 - ₹359,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Description
Availability Pricing
COMPARE
620 100 to 230 Temperature Controller for Fluidized Sand Baths 01184-00 or 01184-02
₹359,000.00INR / Each
COMPARE
620 100 to 230 Temperature Controller for Fluidized Sand Baths 01189-50
₹355,000.00INR / Each