Cole-Parmer® General Purpose Auto Defrost Freezers
Convenient and efficient freezing for general purpose applications
  • Maintenance is hassle-free with an auto defrost system that reduces frost buildup for easy operation
  • Reliable mechanical thermostat with an adjustable range of -15°C to -20°C
  • Secure and practical design with a keyed door lock, recessed handle, probe access port, and a magnetic door gasket for a tight seal
  • Uses natural hydrocarbon refrigerants for a sustainable choice
Cole-Parmer® General Purpose Auto Defrost Freezers provide a straightforward and efficient solution for applications where precise temperature control and maintenance are not required. Featuring a mechanical thermostat, these units offer an adjustable temperature range of -15°C to -20°C, with an automatic defrost system to minimize frost buildup. Designed for convenience, they include security features such as a keyed door lock and a tight-sealing magnetic gasket. While auto defrost cycles may temporarily raise air temperatures, these freezers are a reliable choice for non-critical storage needs. Built with environmentally friendly hydrocarbon refrigerants and backed by a comprehensive warranty, they offer dependable performance for general-purpose applications.
641,000.00 - ₹761,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Min Temperature - Freezer (° C) Power (VAC)
Availability Pricing
COMPARE
14 -25 120
₹641,000.00INR / Each
COMPARE
17 -25 120
₹761,000.00INR / Each