Cole-Parmer® General-Purpose Pipette Tips
Universal fit pipettor tips for educational and general laboratory applications
  • Precision molded from virgin, chemically-resistant and contamination-free, polypropylene (PP)
  • Tested for fit and function with the most widely used pipettes on the market
  • Fully autoclavable in steam at 121°C for 15 minutes.
10,300.00INR / Bag of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Min Volume (mL) Package Of
Availability Pricing
COMPARE
10 1 Bulk; 200/Pk
Call For Price
COMPARE
5 1 Bulk; 2 Bags x 250 Tips/Bag
₹10,300.00INR / Bag of 500