Cole-Parmer® Glass Media Bottles, Class A, Square
Class A square glass media storage bottles ideal for mixing, sampling, and storing mixtures, reagents, solvents, and solutions in the laboratory
  • Resistant to corrosion by most chemicals
  • Withstand temperatures from -50 to 550 °C (-58 to 1022 °F)
  • Wide-mouth opening for easy pouring
  • Autoclavable PP cap and bottle
Made of borosilicate 3.3 glass, these Class A, square media bottles provide outstanding corrosion resistance and meet ASTM E438 requirements. Able to withstand temperatures from -50 to 550 °C (-58 to 1022 °F), they have a wide mouth opening that allows for easy pouring. The space saving, square shape helps you maximize shelf space. Clear, colorless walls have a marking spot for labeling and graduations showing approximate volume. Both the bottle and polypropylene cap are autoclavable.
8,700.00 - ₹33,200.00INR / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Overall Height (cm)
Availability Pricing
COMPARE
100 20 11
₹8,700.00INR / Pkg of 10
COMPARE
250 50 14.5
₹15,300.00INR / Pkg of 10
COMPARE
500 100 18.5
₹20,400.00INR / Pkg of 10
COMPARE
1000 100 23
₹33,200.00INR / Pkg of 10