Cole-Parmer® Glass Stir Rods with Rubber Policemen
Glass stir rods are manufactured with soft borosilicate glass with both ends rounded. They come with a pre-assembled, removable rubber policeman for transferring liquids or clearing precipitate out of containers. Glass rods are chemically resistant and can withstand temperatures to 400°F (205°C) (without rubber policemen). Autoclavable.
3,200.00 - ₹6,100.00INR / Pkg of 12
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material
Availability Pricing
COMPARE
Glass
₹3,200.00INR / Pkg of 12
COMPARE
Glass
₹3,600.00INR / Pkg of 12
COMPARE
Glass
₹3,800.00INR / Pkg of 12
COMPARE
Glass
₹4,300.00INR / Pkg of 12
COMPARE
Glass
₹6,100.00INR / Pkg of 12