Cole-Parmer® Hardened-Ashless Filter Paper
Fast-filtering grade is ideal for gravimetric analysis of coarse particles
  • Suited to collecting wet precipitates
  • Tough, smooth surface free of loose fibers
  • Great for filtering chloride and phosphorus in cement, coal and coke
Use these specially treated hardened-ashless filter papers for critical analytical techniques requiring increased wet-strength and handling capacity. These fast-filtering filter papers are ideal for gelatinous precipitates in acid and alkali solutions, as well as air and food analysis.
4,600.00 - ₹22,200.00INR / Pkg of 100
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm)
Availability Pricing
COMPARE
18.5
₹15,200.00INR / Pkg of 100
COMPARE
12.5
₹8,000.00INR / Pkg of 100
COMPARE
15
₹9,100.00INR / Pkg of 100
COMPARE
11
₹6,600.00INR / Pkg of 100
COMPARE
9
₹5,700.00INR / Pkg of 100
COMPARE
24
₹22,200.00INR / Pkg of 100
COMPARE
4.25
₹4,600.00INR / Pkg of 100
Showing 1- 7 of 7