Cole-Parmer® High Alumina Conical Crucibles
High purity composition for critical analyses
Conical crucibles are 99.7% aluminum oxide. Unglazed inside and out. Maximum temperature is 3182°F (1750°C). Autoclavable.
8,900.00 - ₹22,700.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Material
Availability Pricing
COMPARE
20 High alumina
₹8,900.00INR / Each
COMPARE
30 High alumina
₹12,800.00INR / Each
COMPARE
50 High alumina
₹15,100.00INR / Each
COMPARE
100 High alumina
₹22,700.00INR / Each