Cole-Parmer® HPLC and UHPLC Accessories
Protect your columns from particulate matter to ensure more accurate results and longer service life
  • Maintain chromatographic integrity and increase column lifetime
  • Change out cartridges and filters in seconds reducing down time
Cole-Parmer® guards and filters are designed to ensure that erroneous materials do not find their way into the more important and expensive analytical column.
The direct-connect guard cartridge holder allows for quick replacement of guard cartridges, cartridges sold separately. The guard cartridge system adds sufficient protection to the system without changing retention or selectivity parameters.
An alternative to a guard cartridge system is an in-line filter. These filters have a fingertight direct connection design and are placed between the column and the fitting to trap particulate matter before it reaches the analytical column.
50,600.00 - ₹101,000.00INR / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Pore Size Compatable With Description
Availability Pricing
COMPARE
2 µm Cole-Parmer® HPLC columns HPLC In-Line Filters, 2 µm; 10/Pk
₹101,000.00INR / Pkg of 10
COMPARE
0.5 µm Cole-Parmer® UHPLC columns UHPLC In-Line Filters, 0.5 µm; 4/Pk
₹50,600.00INR / Pkg of 4