Cole-Parmer® KX+ LCGC Certified 2-mL Short Thread Screw Top Glass Vials with Caps
Decrease your risk of failed analysis and sample reruns
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Short thread vials have a wide opening (6.0 mm) with a unique 9-mm thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
Clear glass vials are made of Type I, Class A 33-expansion borosilicate glass. Amber vials are made of Type I, Class B 51-expansion borosilicate glass and are ideal for light-sensitive samples.
6,400.00 - ₹7,800.00INR / Pkg of 100
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Septum Type Special Features Tube Color
Availability Pricing
COMPARE
Rubber/PTFE Label Area Amber
₹6,800.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Standard Amber
₹7,600.00INR / Pkg of 100
COMPARE
Rubber/PTFE Standard Clear
₹6,400.00INR / Pkg of 100
COMPARE
PTFE/Silicone Standard Amber
₹6,900.00INR / Pkg of 100
COMPARE
Rubber/PTFE Standard Amber
₹6,400.00INR / Pkg of 100
COMPARE
Rubber/PTFE Label Area Clear
₹6,500.00INR / Pkg of 100
COMPARE
PTFE/Silicone Label Area Amber
₹7,600.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Label Area Amber
₹7,800.00INR / Pkg of 100
COMPARE
PTFE/silicone Label Area Clear
₹7,300.00INR / Pkg of 100
COMPARE
PTFE/silicone Standard Clear
₹6,600.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Standard Clear
₹7,300.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Label Area Clear
₹7,200.00INR / Pkg of 100
Showing 1- 12 of 12