Cole-Parmer® KX+ LCMS Certified 2-mL Screw Top Glass Vials with Caps
Super clean for highly sensitive GCMS and LCMS applications
  • Vials and caps are lot tested by LCMS for cleanliness—certificate of analysis included, complete with MS scan
  • Minimal pH shift—vials and closures have a neutral pH factor
  • Compatible with all autosamplers
  • Select silicone-free PicoPure Plus septa for MS applications
LCMS certified vials are manufactured using a process that reduces surface activity, improving analytical quantification and minimizing pH shifts in the samples. Surface metals such as sodium and boron that can contaminate samples and interfere with trace analysis are effectively removed. Screw vials feature 9-mm short thread with a unique thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
Clear glass vials are made of Type I, Class A 33-expansion borosilicate glass. Amber vials are made of Type I, Class B 51-expansion borosilicate glass and are ideal for light-sensitive samples.
5,600.00 - ₹7,200.00INR / Pkg of 100
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Septum Type Special Features Tube Color
Availability Pricing
COMPARE
PicoPure Plus silicone-free - Clear
₹5,600.00INR / Pkg of 100
COMPARE
PicoPure Plus silicone-free Label area and filling lines Amber
₹6,300.00INR / Pkg of 100
COMPARE
PicoPure Plus silicone-free - Amber
₹6,000.00INR / Pkg of 100
COMPARE
PicoPure Plus silicone-free Label area and filling lines Clear
₹6,200.00INR / Pkg of 100
COMPARE
PTFE/silicone - Clear
₹6,700.00INR / Pkg of 100
COMPARE
PTFE/silicone - Amber
₹6,700.00INR / Pkg of 100
COMPARE
PTFE/silicone Label area and filling lines Clear
₹6,600.00INR / Pkg of 100
COMPARE
PTFE/silicone Label area and filling lines Amber
₹6,700.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone - Clear
₹6,900.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone - Amber
₹7,200.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Label area and filling lines Clear
₹6,900.00INR / Pkg of 100
Showing 1- 11 of 11