Cole-Parmer® Metal Freezer Boxes
Safely store samples in a liquid nitrogen vapor environment
  • Choose from stainless steel or aluminum
These metal freezer boxes are guaranteed not to rust and feature drain holes making them ideal for storing samples in most liquid nitrogen tanks offered on the market. Available in stainless steel or aluminum, the boxes can be stored in the same rack configurations as the 2” and 3” cardboard boxes. Cardboard dividers can be added depending on the application.
2,800.00 - ₹3,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Metal Freezer Box, Stainless Steel, 2"; 1/Ea
₹3,000.00INR / Each
COMPARE
Metal Freezer Box, Stainless Steel, 3"; 1/Ea
₹3,300.00INR / Each
COMPARE
Metal Freezer Box, Aluminum, 2"; 1/Ea
₹2,800.00INR / Each
COMPARE
Metal Freezer Box, Aluminum, 3"; 1/Ea
₹3,100.00INR / Each