Cole-Parmer® Modular pH Electrode Kits
Preconfigured pH kits make it easy to save; modular design reuses housing and replaces electrode as needed
  • Flat-surface electrode design minimizes electrode damage
  • Self-cleaning feature extends life of electrode
  • Choose from submersible and in-line kits
Purchase a pre-configured kits to meet your application requirements. Economical alternative replaces cartridge and reuses housing and cables.
These self-cleaning, flat surface electrodes have been enhanced with protection against thermal and chemical breakdown. The double-junction pH reference protects from heavy metals, sulfur, iodine, or proteins.
15,200.00 - ₹132,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Cole-Parmer Preconfigured pH electrode Submersible pH/Temperature Kit 2711306
₹132,000.00INR / Each
COMPARE
Preconfigured pH electrode Submersible pH/Temperature Kit
Loading
₹132,000.00INR / Each
Cole-Parmer Preconfigured Modular, Inline pH Kit 2711316
₹130,000.00INR / Each
COMPARE
Preconfigured Modular, Inline pH Kit
Loading
₹130,000.00INR / Each
Cole-Parmer Flow cell, Schedule 80 CPVC, 3/4" x 3/4" FNPT 9967321
₹15,200.00INR / Each
COMPARE
Flow cell, Schedule 80 CPVC, 3/4" x 3/4" FNPT
Loading
₹15,200.00INR / Each