Cole-Parmer® Narrow-Mouth Sample Bottles, Clear Glass
Keep liquid and dry samples safe
  • Made of USP Type III soda-lime glass
  • Chemical resistant PTFE lined cap
  • Keep track of samples with included labels
Manufactured from commercial USP Type III soda-lime glass, each bottle comes with a foam-backed, PTFE cap liner that resists attack from virtually all chemicals at room temperature, ensuring a tight seal to protect liquid samples as well as dry.
Includes a supply of self-adhesive labels.
Caps are not autoclavable.
10,700.00 - ₹17,900.00INR / Case of 48
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
Cole-Parmer 7909 Narrow-Mouth Sample Bottle, Clear Glass, 30mL, 48/CS 3460210
₹17,900.00INR / Case of 48
COMPARE
30 20-400 Glass
Loading
₹17,900.00INR / Case of 48
Cole-Parmer Essentials 7910 Narrow-Mouth Sample Bottle, Clear Glass, 60mL, 24/CS 3460220 Mfr # GLC-01099
₹15,000.00INR / Case of 24
COMPARE
60 20-400 Glass
Loading
₹15,000.00INR / Case of 24
Cole-Parmer Essentials 7912 Narrow-Mouth Sample Bottle, Clear Glass, 125mL, 24/CS 3460230 Mfr # GLC-01123
₹16,800.00INR / Case of 24
COMPARE
120 22-400 Glass
Loading
₹16,800.00INR / Case of 24
Cole-Parmer 7913 Narrow-Mouth Sample Bottle, Clear Glass, 250mL, 24/CS 3460240
₹10,700.00INR / Case of 24
COMPARE
240 24-400 Glass
Loading
₹10,700.00INR / Case of 24
Cole-Parmer 7914 Narrow-Mouth Sample Bottle, Clear Glass, 500mL, 12/CS 3460250
₹14,700.00INR / Case of 12
COMPARE
480 28-400 Glass
Loading
₹14,700.00INR / Case of 12
Cole-Parmer 7917 Narrow-Mouth Sample Bottle, Clear Glass, 1000mL, 12/CS 3460260
₹14,000.00INR / Case of 12
COMPARE
960 33-400 Glass
Loading
₹14,000.00INR / Case of 12