Cole-Parmer® Narrow-Mouth Wash Bottles
No leaking while dispensing
  • Precise delivery with needle-thin stream
These narrow-mouth wash bottles are made of low-density polyethylene (LDPE). One-piece polypropylene (PP) cap/delivery tube helps eliminate leaks at the top. Deliver from a single drop to a needle-thin stream.
3,400.00 - ₹11,000.00INR / Pkg of 12
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
125 28 mm Low Density Polyethyene (LDPE)
₹3,400.00INR / Pkg of 12
COMPARE
250 28 mm Low Density Polyethyene (LDPE)
₹4,000.00INR / Pkg of 12
COMPARE
500 28 mm Low Density Polyethyene (LDPE)
₹11,000.00INR / Pkg of 12
COMPARE
1000 38 mm Low Density Polyethyene (LDPE)
₹9,500.00INR / Pkg of 12