Cole-Parmer® Pellet Mixer Pestles and Microtubes
Quickly homogenize cells, plant tissue, and DNA pellets within microtube vials
  • Free of DNAse, RNAse, and pyrogens to eliminate sample contamination
Plastic pestles and tubes are designed to homogenize cells and plant tissue or to resuspend protein and DNA pellets. They are DNAase/RNAase and pyrogen free, and come individually wrapped to guarantee purity. The pestle features a grooved grip, making it easy to turn without slipping, even while wearing gloves. Pestle length is 3” (7.5 cm). Both pestle and tube are autoclavable. Compatible with model 44468-25 pestle mixer, order separately.
1,600.00 - ₹2,800.00INR / Pkg of 100
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Cole-Parmer Pestle for 1.5 mL Microtube, 100/pk 4446819
₹2,500.00INR / Pkg of 100
COMPARE
Pestle for 1.5 mL Microtube, 100/pk
Loading
₹2,500.00INR / Pkg of 100
Cole-Parmer Pestle for 0.5 mL Microtube, 100/pk 4446820
₹2,400.00INR / Pkg of 100
COMPARE
Pestle for 0.5 mL Microtube, 100/pk
Loading
₹2,400.00INR / Pkg of 100
Cole-Parmer Pellet Mixer Microtube, 1.5 mL; 100/Pk 4446821
₹1,600.00INR / Pkg of 100
COMPARE
Pellet Mixer Microtube, 1.5 mL; 100/Pk
Loading
₹1,600.00INR / Pkg of 100
Cole-Parmer Pestle and 1.5 mL Microtube, 100/pk 4446823
₹2,800.00INR / Pkg of 100
COMPARE
Pestle and 1.5 mL Microtube, 100/pk
Loading
₹2,800.00INR / Pkg of 100
Cole-Parmer Pestle and 0.5 mL Microtube, 100/pk 4446824
₹2,800.00INR / Pkg of 100
COMPARE
Pestle and 0.5 mL Microtube, 100/pk
Loading
₹2,800.00INR / Pkg of 100