Cole-Parmer® PFA Tubing
Superior micro-porosity characteristics and externally smooth finishes—ideal for high-purity applications
  • Made from 100% virgin ultra-high molecular weight perfluoroalkoxy
  • Excellent flexibility compared to other PFA grade tubing
  • Wide temperature range of –160 to 260 °C (–320 to 500 °F)
  • Does not contain BPA, phthalates, or PVC
  • Sterilize by autoclave, EtO, and chemical sterilants
  • Complies with FDA 21 CFR 177.1550, USP Class VI, ASTM D 6867-03
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
The Cole-Parmer PFA tubing, made from ultra-high purity perfluoroalkoxy, is specifically designed to handle harsh chemicals and ultra-high purity applications. Smooth, nonstick surface finish and low permeability result in reduced absorption/adsorption of fluids, minimizing the risk of fluid contamination.
Great flexibility and resistance to cracks make PFA tubing perfect for use in moving parts. High thermal stability characteristics allow use up to 260 °C (500 °F). Translucent tubing makes it easy to view your fluid for sight gauge and flow applications.
13,800.00 - ₹73,500.00INR / Pkg of 1
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/16 1/8 1/32
₹13,800.00INR / Pkg of 1
COMPARE
3/32 5/32 1/32
₹20,200.00INR / Pkg of 1
COMPARE
1/8 3/16 1/32
₹23,300.00INR / Pkg of 1
COMPARE
1/8 1/4 1/16
₹44,700.00INR / Pkg of 1
COMPARE
3/16 1/4 1/32
₹26,800.00INR / Pkg of 1
COMPARE
5/32 1/4 3/64
₹37,000.00INR / Pkg of 1
COMPARE
7/16 1/2 1/32
₹26,800.00INR / Pkg of 1
COMPARE
1/2 9/16 1/32
₹73,500.00INR / Pkg of 1
Showing 1- 8 of 8