Cole-Parmer® pH/Temperature and pH/ORP/Temperature 1/4-DIN Controllers
Isolated and reversible 4 to 20 mA output allow for proportional output, SPDT relays for On/Off control
  • Graphic LCD for clear measurement display
  • Automatic buffer recognition of US and NIST buffers
  • Automatic temperature compensation
  • Password protected ensures security of system
  • RS-485 output for remote access allows multiple unit control from your computer
Cole-Parmer online controllers offer features found in more expensive models. Graphic LCD simultaneously displays measurement, temperature, electrode status, relay status, and transmitter current output. Calibration data is stored in memory; making the unit ready for measurement. Password protection, RS-485 for remote access, and the isolated and reversible 4 to 20 mA output offer extended data options with security.
71,900.00 - ₹111,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
500 pH/Temperature 1/4-DIN Controller
₹92,200.00INR / Each
COMPARE
550 pH/Temperature 1/4-DIN Controller
₹111,000.00INR / Each
COMPARE
500 pH/ORP/Temperature 1/4 DIN Controller
₹71,900.00INR / Each