Cole-Parmer® Pipette Reagent Reservoir
V-shaped angled design maximizes reagent retrieval and reduces waste
  • Certified RNase-, DNase-, and pyrogen-free
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents.
Single Chamber Reservoirs are made of white polystyrene and constructed with low cross bars at each end for added stiffness and strength. Each reservoir features graduations on the inner walls, pour spouts in every corner, and one larger spout to facilitate spill-free pouring. Reservoirs are ideal for 8- and 12-channel pipettors.
12-Chamber Reservoirs are designed for serial dilutions or pipetting different reagents using a 12-channel pipettor. Each numbered chamber holds up to 3 mL. The chemical-resistant polypropylene reservoir can withstand temperatures from -122 to 250°F (-80 to 121°C). Tight fitting lid minimizes evaporation or contamination during incubation or storage.
5,800.00 - ₹13,200.00INR / Case of 50
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Number Of Channels Sterile
Availability Pricing
COMPARE
25 1 Yes
₹5,800.00INR / Case of 50
COMPARE
25 1 Yes
₹10,900.00INR / Case of 100
COMPARE
25 1 No
₹8,600.00INR / Case of 100
COMPARE
50 1 Yes
₹6,800.00INR / Case of 50
COMPARE
50 1 Yes
₹11,600.00INR / Case of 100
COMPARE
50 1 No
₹9,800.00INR / Case of 100
COMPARE
100 1 Yes
₹7,500.00INR / Case of 50
COMPARE
100 1 Yes
₹13,200.00INR / Case of 100
COMPARE
100 1 No
₹10,500.00INR / Case of 100
COMPARE
36 12 No
₹11,700.00INR / Case of 50
COMPARE
36 12 Yes
₹8,800.00INR / Case of 20
Showing 1- 11 of 11