Cole-Parmer® Plus Class A, Type 1 Glass Griffin Low-Form Beakers
Double-scale white graduations for easy reading up and down!
  • Class A borosilicate glass
  • Withstand temperatures from -58°F to 1022°F (-50°C to 550°C)
  • Graduations are durable white enamel
  • Autoclavable
These low-form beakers are constructed of Class A borosilicate glass to meet ASTM E960 specifications.
3,500.00 - ₹12,000.00INR / Pkg of 6
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
10 0 Borosilicate Glass
₹3,500.00INR / Pkg of 12
COMPARE
25 5 Borosilicate Glass
₹4,000.00INR / Pkg of 12
COMPARE
50 10 Borosilicate Glass
₹11,000.00INR / Pkg of 12
COMPARE
100 20 Borosilicate Glass
₹6,000.00INR / Pkg of 12
COMPARE
150 20 Borosilicate Glass
₹6,500.00INR / Pkg of 12
COMPARE
250 50 Borosilicate Glass
₹12,000.00INR / Pkg of 12
COMPARE
400 25 Borosilicate Glass
₹6,500.00INR / Pkg of 8
COMPARE
600 50 Borosilicate Glass
₹12,000.00INR / Pkg of 8
COMPARE
1000 50 Borosilicate Glass
₹10,500.00INR / Pkg of 6
COMPARE
2000 200 Borosilicate Glass
₹8,900.00INR / Pkg of 2
COMPARE
3000 250 Borosilicate Glass
₹5,900.00INR / Each
COMPARE
5000 500 Borosilicate Glass
₹9,000.00INR / Each
Showing 1- 12 of 12