This product is now discontinued, View Related Products
Cole-Parmer® Polystat® Cooling/Heating Circulating Baths
Powerful refrigeration system provides excellent cooling capacity across range of operating temperatures
  • Two-speed pump with up to 21 L/min flow
  • User-friendly interface available in three languages: English, French, German
These baths are ideal for industrial or laboratory applications with a wide variety of temperature control requirements. Available in 3-6 and 8.6-15 liter tank capacities, the compact footprint maximizes bench space. Bright digital display and intuitive interface allow for simple operation. Three languages: English, French, and German.
Robust refrigeration system with metering control provides fast heat removal and stable temperatures. Industrial-grade circulating pumps deliver extra pressure providing more efficient heat exchange, better temperature uniformity, and improved reservoir agitation. Long-life, high-wattage heaters deliver fast fluid heat up for applications requiring time to temperature. High-temperature cutoff and low-level fault protects the bath and application from adverse events. Includes inlet/outlet pump connections for external applications.
490,000.00 - ₹655,000.00INR / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
COMPARE
100 -20 Standard 3-6L Heat/Cool Bath, -20 to 100C; 115VAC
Call For Price
COMPARE
100 -20 Standard 3-6L Heat/Cool Bath, -20 to 100C; 230VAC
₹490,000.00INR / Each
COMPARE
100 -20 Advanced 3-6L Heat/Cool Bath, -20 to 100C; 115VAC
Call For Price
COMPARE
100 -20 Advanced 3-6L Heat/Cool Bath, -20 to 100C; 230VAC
Call For Price
COMPARE
100 -20 Standard 3-6L Heat/Cool Bath, -20 to 100C; 115VAC
Call For Price
COMPARE
100 -20 Standard 3-6L Heat/Cool Bath, -20 to 100C; 230VAC
₹553,000.00INR / Each
COMPARE
100 -20 Advanced 3-6L Heat/Cool Bath, -20 to 100C; 115VAC
Call For Price
COMPARE
100 -20 Advanced 3-6L Heat/Cool Bath, -20 to 100C; 230VAC
Call For Price
COMPARE
100 -28 Standard 8.6-15L Heat/Cool Bath, -28 to 100C; 115VAC
Call For Price
COMPARE
100 -28 Standard 8.6-15L Heat/Cool Bath, -28 to 100C; 230VAC
₹562,000.00INR / Each
COMPARE
150 -28 Standard 8.6L Heat/Cool Bath, -28 to 150C; 115VAC
Call For Price
COMPARE
150 -28 Standard 8.6-15L Heat/Cool Bath, -28 to 150C; 230VAC
₹562,000.00INR / Each
COMPARE
200 -28 Advanced 8.6-15L Heat/Cool Bath, -28 to 200C; 115VAC
Call For Price
COMPARE
200 -28 Advanced 8.6-15L Heat/Cool Bath, -28 to 200C; 230VAC
₹653,000.00INR / Each
COMPARE
100 -35 Standard 8.6-15L Heat/Cool Bath, -35 to 100C; 115VAC
Call For Price
COMPARE
100 -35 Standard 8.6-15L Heat/Cool Bath, -35 to 100C; 230VAC
₹617,000.00INR / Each
COMPARE
150 -35 Standard 8.6-15L Heat/Cool Bath, -35 to 150C; 115VAC
Call For Price
COMPARE
150 -35 Standard 8.6-15L Heat/Cool Bath, -35 to 150C; 230VAC
₹626,000.00INR / Each
COMPARE
200 -35 Advanced 8.6-15L Heat/Cool Bath, -35 to 200C; 115VAC
Call For Price
COMPARE
200 -35 Advanced 8.6-15L Heat/Cool Bath, -35 to 200C; 230VAC
₹655,000.00INR / Each
Showing 1- 20 of 20