Cole-Parmer® Porcelain High-Form Crucibles with Covers
Ashing, drying and concentrating samples now more affordable
  • Withstand temperatures to 1150 °C ( 2102 °F)
  • Autoclavable
These crucibles are glazed on inner and outer surfaces, except outer bottom and rim. Can withstand temperatures to 1150 °C (2102 °F) and heating rates up to 200 °C (392 °F) per hour. The porcelain crucibles provide resistance to acids and alkalies except for hydrofluoric acid.
3,200.00 - ₹79,400.00INR / Pkg of 12
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Height (mm) Capacity (mL) Top Outside Diameter (mm)
Availability Pricing
COMPARE
20 5 24
₹11,800.00INR / Pkg of 12
COMPARE
20 5 24
₹71,100.00INR / Pkg of 72
COMPARE
33 12 27
₹79,400.00INR / Pkg of 72
COMPARE
65 80 51
₹48,800.00INR / Pkg of 24
COMPARE
72 120 59
₹57,500.00INR / Pkg of 24
COMPARE
72 200 62
₹49,100.00INR / Pkg of 12
COMPARE
29 15 35
₹14,300.00INR / Pkg of 12
COMPARE
37 30 43
₹16,900.00INR / Pkg of 12
COMPARE
37 30 43
₹49,900.00INR / Case of 36
COMPARE
40 40 50
₹18,900.00INR / Pkg of 12
COMPARE
40 40 50
₹55,400.00INR / Case of 36
COMPARE
43 50 53
₹21,700.00INR / Pkg of 12
COMPARE
15 1.3 15
₹5,700.00INR / Pkg of 6
COMPARE
20 5 24
₹5,900.00INR / Pkg of 6
COMPARE
26 10 31
₹6,800.00INR / Pkg of 6
COMPARE
29 15 35
₹7,600.00INR / Pkg of 6
COMPARE
32 25 40
₹8,500.00INR / Pkg of 6
COMPARE
37 30 43
₹9,500.00INR / Pkg of 6
COMPARE
40 40 50
₹10,000.00INR / Pkg of 6
COMPARE
43 50 53
₹11,100.00INR / Pkg of 6
Showing 1 - 20 of 25