Cole-Parmer® Porcelain Spatulas with Spoons
Ideal for handling small size samples
  • Resistant to scratches and staining
  • Withstands high temperatures
  • Autoclavable
  • 1 year warranty
These glazed porcelain spatulas with a spoon are durable and long-lasting. They feature one flat and one spoon end for added versatility. The flat end is ideal for scraping, while the spoon end is best used for measuring or transferring small quantities of materials.
Easy to sterilize and can withstand high temperatures.
3,300.00 - ₹11,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Overall Length (cm)
Availability Pricing
COMPARE
0.1 Porcelain 10
₹3,300.00INR / Each
COMPARE
1.2 Porcelain 19.8
₹7,400.00INR / Each
COMPARE
5.5 Porcelain 26
₹11,000.00INR / Each