Cole-Parmer® Precision Titanium Tweezers
Ideal for handling microscopic particles
  • Chemical and corrosion resistant
  • Withstand temperatures up to 800°F (426°C)
These lightweight, nonmagnetic titanium tweezers do not shed particles like stainless steel tweezers and are biocompatible. Typical applications include handling of microscopic particles, sensitive components in cleaning/chemical processes at high temperature, histology, and biology.
9,900.00 - ₹59,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Length (in) Sterile Description
Availability Pricing
COMPARE
4 3/4 No Titanium Tweezers w/ Short, Straight, Fine Tips
₹10,000.00INR / Each
COMPARE
4 3/4 No Titanium Tweezers w/ Very Sharp, Fine Tips
₹9,900.00INR / Each
COMPARE
4 3/4 No Titanium Tweezers w/ Long, Very Sharp, Pointed Tips
₹10,000.00INR / Each
COMPARE
4 1/4 No Titanium Tweezers w/ Short, Very Sharp, Pointed Tips
₹10,000.00INR / Each
COMPARE
4 1/4 No Titanium Tweezers w/ Extra Fine, Polished Tips
₹11,100.00INR / Each
COMPARE
4 1/2 No Titanium Tweezers w/ Curved, Very Fine Tips
₹11,700.00INR / Each
COMPARE
4 1/2 No Precision Titanium Tweezers Kit; 5 Piece Set
₹59,000.00INR / Set
Showing 1- 7 of 7