Cole-Parmer® PVC Fuel and Lubricant Tubing
Specifically formulated to resist hardening and swelling
  • Resist hydrocarbons, avoid swelling, and reduce cracking
  • Safely transfer chemicals and petrochemicals including gasoline, kerosene, oils and cutting compounds
  • Protects against leaching and product contamination
  • Ideal for applications ranging from small engine fuel lines to coolant transfer
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Cole-Parmer® PVC Fuel and Lubricant tubing is the ideal transfer tubing for kerosene, gasoline, oils, cutting compounds, and other chemicals and petrochemicals. This tubing is formulated to resist the corrosive effects of hydrocarbon-based fluids, avoid swelling and hardening, and protect against product contamination and leaching. These fuel line hoses are flexible enough to allow for fast, simple installation in even the tightest areas, and maintain this flexibility over time to reduce failure due to cracking and leaking. This fuel tubing’s transparent yellow color provides for easy identification and flow monitoring, and it is available in the most common sizes to suit nearly any need involving fuel and lubricant applications.
3,100.00 - ₹26,100.00INR / Pkg of 1
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
5/64 9/64 1/32
₹3,100.00INR / Pkg of 1
COMPARE
1/8 1/4 1/16
₹4,500.00INR / Pkg of 1
COMPARE
3/16 5/16 1/16
₹6,500.00INR / Pkg of 1
COMPARE
1/4 3/8 1/16
₹8,000.00INR / Pkg of 1
COMPARE
5/16 7/16 1/16
₹9,500.00INR / Pkg of 1
COMPARE
3/8 1/2 1/16
₹11,400.00INR / Pkg of 1
COMPARE
7/16 9/16 1/16
₹12,800.00INR / Pkg of 1
COMPARE
1/2 3/4 1/8
₹26,100.00INR / Pkg of 1
Showing 1- 8 of 8