Cole-Parmer® Rare Earth Turbo Pivot Stir Bars
Utilize extra-powered samarium-cobalt magnets to provide increased magnetic strength
  • Near totally resistant to demagnetization
  • Identified by a chemically inert carbon black spot
The high magnetic coupling power of samarium-cobalt magnets used in turbo stirrer bars can give rise to erratic stirring effects such as magnet migration and tumbling, while the powerful attraction between the stirrer and the drive magnet system can give rise to the formation of PTFE particles caused by abrasion between stirrer magnet and the container base.
1,300.00 - ₹3,400.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Diameter (mm) Length (mm)
Availability Pricing
COMPARE
3 13
₹1,300.00INR / Each
COMPARE
8 15
₹1,900.00INR / Each
COMPARE
8 25
₹2,600.00INR / Each
COMPARE
8 51
₹3,000.00INR / Each
COMPARE
8 64
₹3,400.00INR / Each