Cole-Parmer® Ratchet-Style Pinch Clamps, Polypropylene
Shut-off the flow with a simple push
  • Lightweight and reusable
  • Rust and corrosion resistant
  • USP Class VI compliant—excellent for biotechnology and pharmaceutical applications for transferring sterile fluids
  • Sterilize by autoclave or gamma irradiation
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Ratchet-style pinch clamps provide an easy way to control the flow in flexible tubing. Multi-position rachet control adjusts to allow flow from full flow to complete stop; simply push down all the way until it latches to stop the fluid flow.
1,700.00 - ₹4,400.00INR / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clamp Material Length (in) Description
Availability Pricing
COMPARE
Polypropylene 1 Ratchet-Style Pinch Clamp, Polypropylene, Small; 25/Pk
₹1,700.00INR / Pkg of 25
COMPARE
Polypropylene 1.5 Ratchet-Style Pinch Clamp, Polypropylene, Medium; 25/Pk
₹2,800.00INR / Pkg of 25
COMPARE
Polypropylene 2.3125 Ratchet-Style Pinch Clamp, Polypropylene, Large; 25/Pk
₹4,400.00INR / Pkg of 25