Cole-Parmer® Rectangular Trays, High-Alumina
Eliminate contamination
  • Highly resistant to chemicals
  • Ideal for high-temperature and high-purity applications
  • Autoclavable
Trays are 99.8% pure aluminum oxide. Highly resistant to the effects of chemicals, these trays are ideal for chemists, metallurgists, and others who are involved in high-temperature work and need contamination-free results.
9,400.00 - ₹144,000.00INR / Case of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (cm) Overall Width (cm)
Availability Pricing
COMPARE
130 15.2 6.5
₹70,100.00INR / Case of 1
COMPARE
750 19.4 14
₹144,000.00INR / Case of 1
COMPARE
5 4 3
₹9,400.00INR / Each
COMPARE
13 6 4
₹12,500.00INR / Each
COMPARE
32 7.5 5
₹14,100.00INR / Each
COMPARE
64 10 4.5
₹28,200.00INR / Each
COMPARE
340 20 9
₹74,900.00INR / Each
Showing 1- 7 of 7