Cole-Parmer® Reinforced Thermoplastic Elastomer (TPE) Tubing
Braided to withstand high-pressure applications
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • Excellent for use in peristaltic pump applications
  • Great alternative to silicone tubing
  • Does not contain PVC, DEHP, phthalates, or other plasticizers
  • Ultra-low extractables and leachables
  • Low gas and oxygen permeability
  • Low protein binding and nonpyrogenic material
  • Thermally weldable
  • USP Class VI, USP 661, and RoHS compliant and BPOG tested
The Cole-Parmer® Reinforced Thermoplastic Elastomer (TPE) tubing is soft and flexible tubing that is durable enough to provide a long service life. This tubing is designed for use with peristaltic pumps. This TPE tubing does not contain PVC, DEHP, phthalates, or plasticizers, making them ideal for various pharmaceutical, food, and beverage applications. The reinforced tubing design makes it ideal for demanding, high pressure use.
11,200.00 - ₹141,000.00INR / Pkg of 1
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
15 5/8 1
₹30,200.00INR / Pkg of 1
COMPARE
25 5/8 1
₹50,300.00INR / Pkg of 1
COMPARE
50 5/8 1
₹100,000.00INR / Pkg of 1
COMPARE
15 3/4 1 1/8
₹42,300.00INR / Pkg of 1
COMPARE
25 3/4 1 1/8
₹70,400.00INR / Pkg of 1
COMPARE
50 3/4 1 1/8
₹141,000.00INR / Pkg of 1
COMPARE
15 1 1 3/8
₹57,600.00INR / Pkg of 1
COMPARE
25 1 1 3/8
₹96,200.00INR / Pkg of 1
COMPARE
15 1/8 3/8
₹11,200.00INR / Pkg of 1
COMPARE
25 1/8 3/8
₹18,500.00INR / Pkg of 1
COMPARE
50 1/8 3/8
₹36,900.00INR / Pkg of 1
COMPARE
15 1/4 1/2
₹11,400.00INR / Pkg of 1
COMPARE
25 1/4 1/2
₹19,000.00INR / Pkg of 1
COMPARE
50 1/4 1/2
₹37,900.00INR / Pkg of 1
COMPARE
15 3/8 5/8
₹17,500.00INR / Pkg of 1
COMPARE
25 3/8 5/8
₹29,300.00INR / Pkg of 1
COMPARE
50 3/8 5/8
₹58,500.00INR / Pkg of 1
COMPARE
25 1/2 3/4
₹35,600.00INR / Pkg of 1
COMPARE
50 1/2 3/4
₹71,000.00INR / Pkg of 1
Showing 1- 19 of 19