This product is now discontinued, View Related Products
Cole-Parmer® Rotational Viscometers
Includes 316 stainless steel spindles – ideal for food applications!
  • Ensure accurate readings by calibrating to a known standard
  • Push-button control with easy-to-use menu system
  • Select from 10 language options
Determine the dynamic viscosity of samples in applications such as food (sauces, juices, and syrups), adhesives, petroleum products, biofuels, paints, pharmaceuticals, chemicals, and more! Over/under range alarm sounds when your sample is too high/low for the spindle selected. All viscometer models feature 10 language options (English, French, German, Italian, Japanese, Portuguese, Spanish, Dutch, Polish, and Catalan), push-button control with easy-to-use menu system, motor self-test, user calibration to a known standard, and universal power supply.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Max Viscosity Range (cP) Output Spindle / Speed Combinations
Availability Pricing
COMPARE
2000000 None 18
Call For Price
COMPARE
13000000 None 18
Call For Price
COMPARE
106000000 None 18
Call For Price
COMPARE
2000000 USB 18
Call For Price
COMPARE
13000000 USB 18
Call For Price
COMPARE
106000000 USB 18
Call For Price
COMPARE
6000000 USB 54
Call For Price
COMPARE
40000000 USB 54
Call For Price
COMPARE
106000000 USB 54
Call For Price
COMPARE
6000000 USB 2600
Call For Price
COMPARE
40000000 USB 2600
Call For Price
COMPARE
106000000 USB 2600
Call For Price
Showing 1- 12 of 12