Cole-Parmer® SamplePrep Vial Cap for Homogenizers
Safely grind samples in the HG-600 Geno/Grinder® 2010 and HG-400 MiniG® homogenizers
  • Screw thread cap ensures a tight seal
These polypropylene caps are lined with a thick 3/32” red silicone disc that prevents cracking. Threaded caps firmly seal Cole-Parmer® homogenizer vials. Replacement caps for 04576-99 and 04575-71. Vials sold separately.
58,100.00INR / Pkg of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
COMPARE
Polyethylene 04576-99 Vials Vial Cap for 15 mL Short Vial, Polyethylene; 100/PK
Call For Price
COMPARE
Polyethylene 04500-38 Vials Vial Cap for 50 mL Vial, Polyethylene; 48/PK
Call For Price
COMPARE
Polypropylene Cap with Silicone Liner 04576-99 and 04575-71 Vials Vial Cap for 15 mL Short Vial, Polypropylene with Silicone Liner; 100/Pk
₹58,100.00INR / Pkg of 100