Cole-Parmer® Sanitary Clamp Fittings, Cross Union, 316 Stainless Steel
Great for food, beverage, and dairy applications—fittings comply with FDA and 3A standards
  • Ideal for general-purpose, food, beverage, and pharmaceutical applications
  • Free technical application support available to assist with tubing selection
  • 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases
  • Easily combine or split fluid flow
Use these 316L stainless steel sanitary fittings in the food, beverage, and pharmaceutical industries. They provide excellent chemical compatibility and corrosion resistance while offering a leak-proof connection; use with our SS clamp for a secure seal. Easily remove to clean or use Clean-in-Place (CIP) process to reduce the chances of dirt and bacteria contaminating the process.
11,000.00 - ₹15,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Number Of Ports
Availability Pricing
COMPARE
316L Stainless Steel 1 4
₹11,000.00INR / Each
COMPARE
316L Stainless Steel 1-1/2 4
₹11,900.00INR / Each
COMPARE
316L Stainless Steel 2 4
₹15,300.00INR / Each