Cole-Parmer® Sanitary Clamp Fittings, Straight Eccentric Reducer, 316 Stainless Steel
Great for food, beverage, and dairy applications—fittings comply with FDA and 3A standards
  • Ideal for general-purpose, food, beverage, and pharmaceutical applications
  • Free technical application support available to assist with tubing selection
  • 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases
  • Offset centerline—useful when both air and liquids are flowing together
Use these 316L stainless steel sanitary fittings in the food, beverage, and pharmaceutical industries. They provide excellent chemical compatibility and corrosion resistance while offering a leak-proof connection; use with our SS clamp for a secure seal. Easily remove to clean or use Clean-in-Place (CIP) process to reduce the chances of dirt and bacteria contaminating the process.
6,300.00 - ₹10,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Sanitary Clamp Size (Port 2) (in)
Availability Pricing
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Eccentric Sanitary Clamp Reducer, 1" x 3/4" 5011804
₹6,300.00INR / Each
COMPARE
316L Stainless Steel 1 3/4
Ships by April 11
₹6,300.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Eccentric Sanitary Clamp Reducer, 1-1/2" x 1" 5011805
₹6,800.00INR / Each
COMPARE
316L Stainless Steel 1-1/2 1
Ships by April 11
₹6,800.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Eccentric Sanitary Clamp Reducer, 2" x 1" 5011806
₹10,400.00INR / Each
COMPARE
316L Stainless Steel 2 1
Ships by April 11
₹10,400.00INR / Each