Cole-Parmer® Sanitary Clamp Fittings, Straight Threaded Adapter, 316 Stainless Steel
Great for food, beverage, and dairy applications—fittings comply with FDA and 3A standards
  • Ideal for general-purpose, food, beverage, and pharmaceutical applications
  • Free technical application support available to assist with tubing selection
  • 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases
  • Connect to pipes using the NPT(M) thread connection
Use these 316L stainless steel sanitary fittings in the food, beverage, and pharmaceutical industries. They provide excellent chemical compatibility and corrosion resistance while offering a leak-proof connection; use with our SS clamp for a secure seal. Easily remove to clean or use Clean-in-Place (CIP) process to reduce the chances of dirt and bacteria contaminating the process.
4,600.00 - ₹7,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Thread Size NPT(M) (Port 2) (InchShort)
Availability Pricing
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 3/4" Clamp x 1/8" NPT(M) 5011815
₹4,600.00INR / Each
COMPARE
316L Stainless Steel 3/4 1/8
Loading
₹4,600.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 3/4" Clamp x 1/4" NPT(M) 5011816
₹4,600.00INR / Each
COMPARE
316L Stainless Steel 3/4 1/4
Loading
₹4,600.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 3/4" Clamp x 3/8" NPT(M) 5011817
₹4,600.00INR / Each
COMPARE
316L Stainless Steel 3/4 3/8
Loading
₹4,600.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 3/4" Clamp x 1/2" NPT(M) 5011818
₹4,600.00INR / Each
COMPARE
316L Stainless Steel 3/4 1/2
Loading
₹4,600.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 3/4" Clamp x 3/4" NPT(M) 5011819
₹4,900.00INR / Each
COMPARE
316L Stainless Steel 3/4 3/4
Loading
₹4,900.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 1" Clamp x 3/4" NPT(M) 5011820
₹5,900.00INR / Each
COMPARE
316L Stainless Steel 1 3/4
Loading
₹5,900.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 1" Clamp x 1" NPT(M) 5011821
₹6,200.00INR / Each
COMPARE
316L Stainless Steel 1 1
Loading
₹6,200.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 1" Clamp x 1-1/2" NPT(M) 5011822
₹7,000.00INR / Each
COMPARE
316L Stainless Steel 1 1 1/2
Loading
₹7,000.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 1-1/2" Clamp x 3/4" NPT(M) 5011823
₹6,100.00INR / Each
COMPARE
316L Stainless Steel 1-1/2 3/4
Loading
₹6,100.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 1-1/2" Clamp x 1" NPT(M) 5011824
₹6,200.00INR / Each
COMPARE
316L Stainless Steel 1-1/2 1
Loading
₹6,200.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 1-1/2" Clamp x 1-1/4" NPT(M) 5011825
₹6,400.00INR / Each
COMPARE
316L Stainless Steel 1-1/2 1 1/4
Loading
₹6,400.00INR / Each
Cole-Parmer Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 1-1/2" Clamp x 1-1/2" NPT(M) 5011826
₹7,000.00INR / Each
COMPARE
316L Stainless Steel 1-1/2 1 1/2
Loading
₹7,000.00INR / Each
Showing 1- 12 of 12