Cole-Parmer® Sanitary Clamp Fittings, Tee Union, 316 Stainless Steel
Great for food, beverage, and dairy applications—fittings comply with FDA and 3A standards
  • Ideal for general-purpose, food, beverage, and pharmaceutical applications
  • Free technical application support available to assist with tubing selection
  • 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases
  • Easily combine or split fluid flow
Use these 316L stainless steel sanitary fittings in the food, beverage, and pharmaceutical industries. They provide excellent chemical compatibility and corrosion resistance while offering a leak-proof connection; use with our SS clamp for a secure seal. Easily remove to clean or use Clean-in-Place (CIP) process to reduce the chances of dirt and bacteria contaminating the process.
8,600.00 - ₹12,200.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Number Of Ports
Availability Pricing
COMPARE
316L Stainless Steel 3/4 3
₹8,600.00INR / Each
COMPARE
316L Stainless Steel 1 3
₹9,100.00INR / Each
COMPARE
316L Stainless Steel 1-1/2 3
₹9,600.00INR / Each
COMPARE
316L Stainless Steel 2 3
₹12,200.00INR / Each