Cole-Parmer® Sanitary Fittings, Tee Union, Polypropylene
High-quality nonmetallic sanitary fittings for critical fluid applications
  • Meet US and EU standards for use in pharmaceutical process applications
  • ASME-BPE, USP Class VI, and FDA compliant
  • Sterilize by autoclave, EtO, or gamma irradiation
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Use tee unions to combine or split fluid flow. These single-use polypropylene sanitary fittings are ideal for applications where metal is incompatible. Smooth inner surface and seamless construction prevent particle entrapment, eliminating potential bacterial growth. Sanitary fittings are great for cell culture media, aseptic fill and transfer, and sterile filling/emptying applications. Order a clamp and gasket to complete your system.
7,200.00 - ₹8,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Connection Type Sanitary Clamp Size (Port 1) (in)
Availability Pricing
COMPARE
Polypropylene Sanitary 3/4
₹7,200.00INR / Each
COMPARE
Polypropylene Sanitary 1
₹8,800.00INR / Each
COMPARE
Polypropylene Sanitary 1-1/2
₹8,600.00INR / Each