Cole-Parmer® Sanitary Gaskets, PTFE
Compatible with Tri-Clamp® style fittings
  • Offers excellent chemical and heat resistance
  • Exhibits great stability
  • Low absorption rate and extractable values
  • Usable temperature range: –73 to 260 °C (–100 to 500 °F)
  • Animal-derived ingredient free and phthalate free
  • FDA CFR 21 177 2600, USP Class VI, and 3A compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
These PTFE gaskets offer excellent chemical, heat, stress, and corrosion resistance for worry-free and leak-free fluid transfer. Gaskets are free of phthalates, TSE/BSE, and animal-derived ingredients, ensuring the integrity of your fluid. Additionally, PTFE gaskets also comply with CFR Title 21: 177.1550 and meet the general requirements of the current 3A sanitary standards.
2,900.00 - ₹3,900.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sanitary Gasket Size (in) Seal Material Description
Availability Pricing
COMPARE
1/2 PTFE Sanitary Gasket, PTFE, 1/2"; 10/Pk
₹2,900.00INR / Pkg of 10
COMPARE
3/4 PTFE Sanitary Gasket, PTFE, 3/4"; 10/Pk
₹2,900.00INR / Pkg of 10
COMPARE
1 PTFE Sanitary Gasket, PTFE, 1"; 10/Pk
₹3,800.00INR / Pkg of 10
COMPARE
1 1/2 PTFE Sanitary Gasket, PTFE, 1-1/2"; 10/Pk
₹3,000.00INR / Pkg of 10
COMPARE
2 PTFE Sanitary Gasket, PTFE, 2"; 10/Pk
₹3,500.00INR / Pkg of 10
COMPARE
2 1/2 PTFE Sanitary Gasket, PTFE, 2-1/2"; 10/Pk
₹3,900.00INR / Pkg of 10