Cole-Parmer® Scrapers, HDPE
Easily remove stuck on residue
  • Constructed from durable high-density polyethylene (HDPE)
  • Cleanroom class 100,000 produced and individually packaged
  • Disposable scrapers are ideal for food and pharmaceutical applications
These scrapers are ideal for food, creams, and general scaping. Disposable scrapers are molded and packaged in class 100,000 cleanroom and individually bagged to avoid cross contamination.
79,300.00 - ₹102,000.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Length (in) Sterile Width (in)
Availability Pricing
COMPARE
7 1/2 No 3
₹79,300.00INR / Pkg of 100
COMPARE
7 1/2 Yes 3
₹102,000.00INR / Pkg of 100
COMPARE
7 1/2 No 4 19/64
₹93,300.00INR / Pkg of 100
COMPARE
7 1/2 Yes 4 19/64
₹99,900.00INR / Pkg of 100