Cole-Parmer® Shell Vials with Plugs
Vials and caps in a convenient pack of 100
  • (PE) plugs give a tight seal yet are easy to insert or remove
  • Fits into bench drawer for easy access.
Components packaged in a clear lid tray with 100 shell vials and 100 conical snap plugs. Snap plugs are not attached to the vials and are in individual plastic bags. Separate packaging keeps vials and plugs particle free.
4,100.00 - ₹7,300.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Diameter (mm) Tube Material Volume (mL)
Availability Pricing
COMPARE
8 Polypropylene 0.7
₹6,800.00INR / Pkg of 100
COMPARE
8 Borosilicate Glass 1
₹4,100.00INR / Pkg of 100
COMPARE
12 Borosilicate Glass 2
₹6,700.00INR / Pkg of 100
COMPARE
15 Borosilicate Glass 4
₹7,300.00INR / Pkg of 100