Cole-Parmer® Short Thread Glass Vials, 9 mm
Compatible with all autosamplers
  • Made from 1st hydrolytic class 51 expansion glass
Short thread vials have a wide opening (6.0 mm ID) with a unique thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
Order silanized vials to minimize adsorption of polar compounds, including amino acids, proteins, and phenols to the inner surface of the vial. High-recovery vials deliver high sample capacity with very low residual volume. Use total-recovery vials for maximum sample recovery.
17,400.00 - ₹370,000.00INR / Pkg of 1000
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Special Features Tube Color Volume (mL)
Availability Pricing
COMPARE
- Clear 2
₹17,400.00INR / Pkg of 1000
COMPARE
- Amber 2
₹23,500.00INR / Pkg of 1000
COMPARE
Label and filling lines Clear 2
₹18,800.00INR / Pkg of 1000
COMPARE
Label and filling lines Amber 2
₹20,200.00INR / Pkg of 1000
COMPARE
Silanized, label, and filling lines Amber 2
₹82,800.00INR / Pkg of 1000
COMPARE
Silanized Clear 2
₹75,000.00INR / Pkg of 1000
COMPARE
Fused insert and label Clear 0.35
₹177,000.00INR / Pkg of 1000
COMPARE
Champagne glass bottom Clear 1.5
₹135,000.00INR / Pkg of 1000
COMPARE
Champagne glass bottom Amber 1.5
₹131,000.00INR / Pkg of 1000
COMPARE
Base bonded insert Clear 2
₹120,000.00INR / Pkg of 1000
COMPARE
Base bonded insert Amber 2
₹134,000.00INR / Pkg of 1000
COMPARE
High recovery Clear 1.1
₹122,000.00INR / Pkg of 1000
COMPARE
Silanized, high recovery Clear 1.5
₹201,000.00INR / Pkg of 1000
COMPARE
Total recovery Clear 0.9
₹370,000.00INR / Pkg of 1000
Showing 1- 14 of 14