Cole-Parmer® Snap Top Vial Smart Packs
Smart Packs combine vials and snap caps for your convenience
  • Resealable box helps prevent contamination during use
  • Select from various septa material, thickness, and hardness
Snap ring vial smart packs come with 2-mL glass vials plus 11 mm hard polyethylene (PE) snap caps with septum.
50,400.00 - ₹78,900.00INR / Pkg of 1000
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Septum Type Special Features Tube Color
Availability Pricing
COMPARE
Natural rubber/transparent PTFE; 1.0 mm; 60° shore A Wide opening Clear
₹50,400.00INR / Pkg of 1000
COMPARE
Red PTFE/white silicone/red PTFE; 1.0 mm; 45° shore A Wide opening Clear
₹78,900.00INR / Pkg of 1000
COMPARE
UltraClean™ white silicone/red PTFE; 1.3 mm; 45° shore A Wide opening Clear
₹66,200.00INR / Pkg of 1000