Cole-Parmer® StableTemp™ Laboratory Refrigerators
Safe cold storage in convenient compact sizes
  • Forced-air cooling with internal fan for maximum temperature uniformity and stability
  • Automatic defrost maintains optimum cooling capacity
  • Multiple alarm system for safe and reliable operation
  • Compressor warranty: 3 years
Designed to meet the demanding requirements of everyday use, these upright refrigerators with microprocessor controller provide energy-efficient, safe, and reliable storage of temperature-sensitive items in a variety of applications. Complete temperature alarm system features an audible buzzer and visible flashing light that will alert for failure due to high or low temperature, sensor error, door ajar, low battery, or power failure. Available with safety lock to prevent unauthorized access.
Call For Price
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Power (Hz) Power (VAC)
Availability Pricing
COMPARE
2.4 50 230
Call For Price
COMPARE
2.4 50 230
Call For Price
COMPARE
4.2 50 230
Call For Price
COMPARE
4.2 50 230
Call For Price
COMPARE
10.2 50 230
Call For Price
COMPARE
13.8 50 230
Call For Price
COMPARE
21.6 50 230
Call For Price
COMPARE
31.4 50 230
Call For Price
COMPARE
48.7 50 230
Call For Price
Showing 1- 9 of 9