Cole-Parmer® StableTemp™ Spark Free Freezers
Designed for safe storage of samples with explosion-proof interior with grounded metal parts
  • Explosion-proof interior
  • EU explosion proof ATEX II C-T6
  • HC System to save energy
  • Micro-processor control system with LCD
  • Dual-lock design
  • Warranty: 1 year on main unit, 3 years on compressor
Cole-Parmer StableTemp Spark Free series is microprocessor controlled with LCD temperature display and adjustable range of -30°C to -10°C (-25°C to -10°C) to for 3.3 Cu Ft model); 0.1°C increments. Optimized hydrocarbon refrigeration system design ensures a more energy efficient operation, meets EU RoHS directive for a green and safe environment.
136,000.00 - ₹350,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (Hz) Power (VAC) Description
Availability Pricing
COMPARE
50 240 Spark Free Upright Freezer, 9.8 CuFt, -30°C, 220-240 VAC, 50 Hz; 1/Ea
₹350,000.00INR / Each
COMPARE
50 240 Spark Free Upright Freezer, 3.3 CuFt, -30°C, 220-240 VAC, 50 Hz; 1/Ea
₹136,000.00INR / Each