Cole-Parmer® Stainless Steel Tweezers with Ceramic Tips
Ceramic tips offer high temperature stability
  • Corrosion resistant and inert to most chemicals, salts, and acids
These antimagnetic, anti-acid stainless steel body tweezers with replaceable ceramic precision tips are ideal where corrosion resistance and toughness are primary requirements. Made from zirconia, the ceramic tips offer superior strength, flexibility, and thermal properties. The tips can be quickly and easily replaced and feature a unique tip alignment. Typical applications include the electronic industry, watch makers, jewelers, and laboratory applications.
16,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Length (cm) Overall Length (in) Style
Availability Pricing
COMPARE
14 5 33/64 Flat Duckbill Tip
₹16,300.00INR / Each
COMPARE
13.5 5 5/16 Fine Curved Tip
₹16,300.00INR / Each
COMPARE
13.5 5 5/16 Fine Thin Tip
₹16,300.00INR / Each
COMPARE
14 5 33/64 Fine Point Tip
₹16,300.00INR / Each