Cole-Parmer® Standard Series Laboratory Refrigerators
Keep your critical samples safe with precise temperature control
  • Precise temperature management with a digital microprocessor controller that ensures accurate control from 1°C to 10°C
  • Forced air directional system maintains uniformity and rapid recovery after door openings
  • Built-in safety features such as audible/visual alarms, resettable Min/Max, and a keyed door lock for added security
  • Uses natural hydrocarbon refrigerants for an environmentally responsible choice
Cole-Parmer® Standard Series Laboratory Refrigerators are designed to meet the stringent demands of laboratory environments. Featuring a state-of-the-art digital microprocessor temperature controller, these refrigerators provide exceptional temperature precision to protect your critical samples and supplies. The advanced forced air directional refrigeration system ensures consistent temperature uniformity and rapid recovery, even with frequent door openings. Safety and convenience are built-in, with features like a cycle-defrost system to prevent damaging temperature spikes, remote alarm contacts, and a secure magnetic door seal. Choose reliability and peace of mind with a unit that combines advanced technology, durability, and eco-conscious design.
1,030,000.00 - ₹1,756,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Doors Power (VAC) Refrigerator Capacity (cu ft)
Availability Pricing
Cole-Parmer Standard Refrigerator, Glass Door, 1°C to 10°C, 16 cubic ft, 120V 4426200
₹1,030,000.00INR / Each
COMPARE
1 glass 120 16
Loading
₹1,030,000.00INR / Each
Cole-Parmer Standard Refrigerator, Glass Door, 1°C to 10°C, 26 cubic ft, 120V 4426201
₹1,269,000.00INR / Each
COMPARE
1 glass 120 26
Loading
₹1,269,000.00INR / Each
Cole-Parmer Standard Refrigerator, 2 Glass Doors, 1°C to 10°C, 49 cubic ft, 120V 4426202
₹1,756,000.00INR / Each
COMPARE
2 glass 120 49
Loading
₹1,756,000.00INR / Each
Cole-Parmer Standard Refrigerator, Solid Door, 1°C to 10°C, 26 cubic ft, 120V 4426203
₹1,269,000.00INR / Each
COMPARE
1 solid 120 26
Loading
₹1,269,000.00INR / Each
Cole-Parmer Standard Refrigerator, 2 Solid Doors, 1°C to 10°C, 49 cubic ft, 120V 4426204
₹1,756,000.00INR / Each
COMPARE
2 solid 120 49
Loading
₹1,756,000.00INR / Each
Cole-Parmer Standard Refrigerator, Solid Door, 1°C to 10°C, 14 cubic ft, 120V 4426217
₹1,049,000.00INR / Each
COMPARE
1 solid 120 14
Loading
₹1,049,000.00INR / Each
Cole-Parmer Standard Refrigerator, Solid Door, 1°C to 10°C, 17 cubic ft, 120V 4426218
₹1,117,000.00INR / Each
COMPARE
1 solid 120 17
Loading
₹1,117,000.00INR / Each
Cole-Parmer Standard Refrigerator, Solid Door, 1°C to 10°C, 20 cubic ft, 120V 4426219
₹1,175,000.00INR / Each
COMPARE
1 solid 120 20
Loading
₹1,175,000.00INR / Each
Showing 1- 8 of 8