Cole-Parmer® TELOS® C18 Nonpolar SPE Columns
Extract hydrophobic or polar organic analytes from aqueous matrices
  • Features C18 sorbent
TELOS nonpolar (reversed phase) SPE columns are available in a wide range of sorbents for extraction of hydrophobic and polar analytes. Select columns with endcaps for applications requiring minimal secondary silanol interactions to enhance compound retention and elution. All columns are made of polypropylene and feature Luer-slip outlet.
Column 06476-61 comes prefitted with ASPEC Dec cap/adapter. TELOS C18 has a pore size of 70 Å (except 06476-71 with 120 Å and 06476-97 with 60 Å) and average particle size of 50 µm (except 06476-91 with 130 to 150 µm).
8,200.00 - ₹20,900.00INR / Pkg of 30
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg) End Cap
Availability Pricing
COMPARE
6 1000 No
₹12,600.00INR / Pkg of 30
COMPARE
15 2000 No
₹15,700.00INR / Pkg of 20
COMPARE
1 100 No
₹19,600.00INR / Pkg of 100
COMPARE
1 100 No
₹20,900.00INR / Pkg of 100
COMPARE
3 100 No
₹13,100.00INR / Pkg of 50
COMPARE
3 200 No
₹14,600.00INR / Pkg of 50
COMPARE
10 200 No
₹8,500.00INR / Pkg of 20
COMPARE
3 500 No
₹13,100.00INR / Pkg of 50
COMPARE
6 500 No
₹9,200.00INR / Pkg of 30
COMPARE
10 500 No
₹8,200.00INR / Pkg of 20
COMPARE
6 1000 Yes
₹13,900.00INR / Pkg of 30
COMPARE
15 2000 Yes
₹15,700.00INR / Pkg of 20
COMPARE
1 50 Yes
₹18,800.00INR / Pkg of 100
COMPARE
1 100 Yes
₹19,600.00INR / Pkg of 100
COMPARE
3 200 Yes
₹14,800.00INR / Pkg of 50
COMPARE
3 500 Yes
₹12,900.00INR / Pkg of 50
COMPARE
6 500 Yes
₹9,200.00INR / Pkg of 30
COMPARE
10 500 Yes
₹8,500.00INR / Pkg of 20
Showing 1- 18 of 18