Cole-Parmer® TELOS® Supported Liquid Extraction (SLE) Plates
Perfectly suited for automation and high throughput procedures
  • Efficient extraction
  • High analyte recovery with no emulsion formation
Use supported liquid extraction plates when you need to extract samples quickly and effectively. Wells are packed with a diatomaceous earth, a neutral matrix, to provide support to absorb the aqueous sample. Once the aqueous sample is fully absorbed, a water immiscible organic solvent is used to elute the analytes. High analyte recoveries are obtained without any offline steps such as protein precipitation, and sample extracts are free of proteins and phospholipids. A hydrophilic top frit allows the sample to pass through while minimizing any blockage.
36,400.00 - ₹65,600.00INR / Pkg of 100
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Applications Bed Mass (mg) Description
Availability Pricing
COMPARE
Neutral matrix (NM) 545E 50 SLE Supported Liquid Extraction Wells, Neutral Matrix 545E, 50 mg; 100/PK
₹45,900.00INR / Pkg of 100
COMPARE
Neutral matrix (NM) 545E 50 SLE Supported Liquid Extraction Plate, 96-Well, Neutral Matrix 545E, 50 mg; 1/EA
₹44,500.00INR / Each
COMPARE
Neutral matrix 400 SLE Supported Liquid Extraction Plate, 96-Well, Neutral Matrix pH 7, 400 mg; 1/EA
₹50,100.00INR / Each
COMPARE
Neutral matrix 100 SLE Supported Liquid Extraction Plate, 96-Well, Neutral Matrix pH 9, 100 mg; 1/EA
₹36,400.00INR / Each
COMPARE
Neutral matrix 200 SLE Supported Liquid Extraction Plate, 96-Well, Neutral Matrix pH 9, 200 mg; 1/EA
₹44,500.00INR / Each
COMPARE
Neutral matrix 600 SLE Supported Liquid Extraction Plate, 96-Well, Neutral Matrix pH 7, 600 mg; 1/EA
₹54,800.00INR / Each
COMPARE
Neutral matrix 600 SLE Supported Liquid Extraction Plate, 96-Well, Neutral Matrix pH 9, 600 mg; 1/EA
₹65,600.00INR / Each
COMPARE
Neutral matrix 200 SLE Supported Liquid Extraction Plate, 96-Well, Neutral Matrix pH 7, 200 mg; 1/EA
₹46,300.00INR / Each
COMPARE
Neutral matrix 400 SLE Supported Liquid Extraction Plate, 96-Well, Neutral Matrix pH 9, 400 mg; 1/EA
₹48,200.00INR / Each
Showing 1- 9 of 9