Cole-Parmer® Thermocouple Hypodermic Tip Penetration Probes with Handle
Hypodermic Tip Penetration Probes Are Designed for Soft and Semi-Soft Materials
  • Ideal for cecking food temperatures
  • Response Time: 1.2 seconds or greater
  • ABS handles provide a light weight design for ease of use along with durability.
  • Probe handles feature a strain relief spring that protects the cable connection against damage due to repeated flexing and tugging.
  • Each probe comes with color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue.
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
13,500.00 - ₹18,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
400 J
₹13,500.00INR / Each
COMPARE
600 K
₹13,500.00INR / Each
COMPARE
400 T
₹18,400.00INR / Each