Cole-Parmer® Tubing Cutters
Durable steel blade makes straight, clean cuts through plastic tubing
  • Steel blade offers durability
Use tubing cutters to make clean, straight cuts through your plastic tubing. Straight cuts ensure better connection to fittings.
530.00 - ₹1,600.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Color Compatible With Description
Availability Pricing
COMPARE
Blue Tubing with 1/16" to 5/8" OD Benchtop Tubing Cutter, 1/16" to 5/8" OD
₹1,600.00INR / Each
COMPARE
Silver Benchtop tubing cutter 50118-48 Replacement Blade for Benchtop Tubing Cutter
₹530.00INR / Each
COMPARE
Black Work with tubing up to 1-1/4" OD (1-1/2" OD for PE tubing) Handheld Tubing Cutter, PTFE Coated Steel, Max 1-1/4 OD Tubing
₹1,600.00INR / Each